missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetraethylene Glycol Diacrylate (stabilized with MEHQ), TCI America™

Manufacturer:  TCI America T156925G

 View more versions of this product

Catalog No. T156925G



Specifications

Specifications

Tetraethylene Glycol Diacrylate (stabilized with MEHQ)
25g
C14H22O7
1760
C=CC(=O)OCCOCCOCCOCCOC(=O)C=C
302.323
302.32
≥90.0% (GC)
Yellow
17831-71-9
MFCD00008630
HCLJOFJIQIJXHS-UHFFFAOYSA-N
2-[2-[2-(2-prop-2-enoyloxyethoxy)ethoxy]ethoxy]ethyl prop-2-enoate
28803
Clear Liquid at 20°C

Chemical Identifiers

17831-71-9
302.323
HCLJOFJIQIJXHS-UHFFFAOYSA-N
2-[2-[2-(2-prop-2-enoyloxyethoxy)ethoxy]ethoxy]ethyl prop-2-enoate
C14H22O7
MFCD00008630
28803
C=CC(=O)OCCOCCOCCOCCOC(=O)C=C
Safety and Handling

Safety and Handling

EINECSNumber : (7)-0152

RTECSNumber : AS8100000

TSCA : Yes

Documents