Please login to your online account to display your discounted pricing

Tetraethylemethylenediphosphonate, 97%, ACROS Organics™


Quantity 1mL

Manufacturer: thermo fisher scientific  AC308820010

 View more versions of this product

Catalog No. AC308820010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 288.25
Assay ≥96.0%
Physical Form Liquid
Quantity 1mL
Color Clear yellow to yellow-brown
Density 1.1600g/mL
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay Percent Range 97%
Molecular Formula C9H22O6P2
CAS 1660-94-2
Chemical Name or Material Tetraethylemethylenediphosphonate
MDL Number MFCD00039887
Infrared Spectrum Authentic
Refractive Index 1.4405 to 1.4445 (20°C, 589nm)
Linear Formula CH[P(O)(OCH5)]

Specifications

Chemical Name or Material Tetraethylemethylenediphosphonate
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H22O6P2
Linear Formula CH[P(O)(OCH5)]
Formula Weight 288.25
MDL Number MFCD00039887
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents