missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetraethylammonium tetrafluoroborate, 99%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 221611000

 View more versions of this product

Catalog No. AC221611000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Tetraethylammonium tetrafluoroborate
100g
98.5% min.
Glass bottle
Crystals or Crystalline Powder
C8H20N·BF4
217.06
White
429-06-1
Authentic
Solubility in water: Soluble
99%
(C2H5)4NBF4
MFCD00011827

Chemical Identifiers

99%
(C2H5)4NBF4
MFCD00011827
C8H20N·BF4
217.06
Tetraethylammonium tetrafluoroborate

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.