missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraethylammonium tetrafluoroborate, 99%

Manufacturer:  Alfa Aesar™ A1021130

 View more versions of this product

Catalog No. AAA1021130



Description

Description

Applications
Tetraethylammonium tetrafluoroborate is used as a phase transfer catalyst in organic synthesis. It finds application as an electrolyte additive in the capillary electrophoretic separation of polyphenols found in grape seed extracts. It is also used to prepare other tetrabutylammonium salts in aqueous solutions. It is involved in the synthesis of conducting poly(thiophenes).

Solubility
Soluble in water, alcohol and acetonitrile.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications

Specifications

Tetraethylammonium tetrafluoroborate
250g
C8H20BF4N
MFCD00011827
3917638
XJRAKUDXACGCHA-UHFFFAOYSA-N
tetraethylazanium;tetrafluoroborate
2724277
99%
370°C to 380°C (decomposition)
429-06-1
(CH3CH2)4NBF4
UN2811
tetraethylammonium tetrafluoroborate, tetraethylazanium tetrafluoroborate, tetraethylammoniumtetrafluoroborate, acmc-1ago3, ksc238k1t, ethanaminium, n,n,n-triethyl-, tetrafluoroborate 1-, ethanaminium, n,n,n-triethyl-, tetrafluoroborate 1-1:1, tetraethyl ammonium tetrafluoroborate, tetraethylammonium tetra fluoro borate
[B-](F)(F)(F)F.CC[N+](CC)(CC)CC
217.059
217.06

Chemical Identifiers

429-06-1
217.059
XJRAKUDXACGCHA-UHFFFAOYSA-N
2724277
[B-](F)(F)(F)F.CC[N+](CC)(CC)CC
C8H20BF4N
MFCD00011827
tetraethylammonium tetrafluoroborate, tetraethylazanium tetrafluoroborate, tetraethylammoniumtetrafluoroborate, acmc-1ago3, ksc238k1t, ethanaminium, n,n,n-triethyl-, tetrafluoroborate 1-, ethanaminium, n,n,n-triethyl-, tetrafluoroborate 1-1:1, tetraethyl ammonium tetrafluoroborate, tetraethylammonium tetra fluoro borate
tetraethylazanium;tetrafluoroborate
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : TSCA

SDS Documents