missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraethylammonium perchlorate, 98% (dry wt.), cont. ≈10% water

Manufacturer:  Alfa Aesar™ 4406218

 View more versions of this product

Catalog No. AA4406218



Specifications

Specifications

Tetraethylammonium perchlorate
>300°C
2567-83-1
C8H20ClNO4
UN1479
acmc-1ccq7
Soluble in water,acetone,acetonitrile and dimethyl sulfoxide.
CC[N+](CC)(CC)CC.[O-]Cl(=O)(=O)=O
229.701
229.71
98%
Odorless
50g
(dry wt.)
MFCD00041977
3638093
Contains ≈ 10% water
WGHUNMFFLAMBJD-UHFFFAOYSA-M
tetraethylazanium;perchlorate
2734932
Wet Crystals

Chemical Identifiers

2567-83-1
229.701
WGHUNMFFLAMBJD-UHFFFAOYSA-M
2734932
CC[N+](CC)(CC)CC.[O-]Cl(=O)(=O)=O
C8H20ClNO4
MFCD00041977
acmc-1ccq7
tetraethylazanium;perchlorate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P271-P280-P283-P302+P352-P304+P340-P305+P351+P338-P306+P360-P312-P332+P313-P363-P370+P378q-P371+P380+P375-P501c

H271-H315-H319-H335

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.

EINECSNumber : 219-904-3

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents