Please login to your online account to display your discounted pricing

Tetraethylammonium perchlorate, 0.2M aqueous solution, ACROS Organics™


Quantity 500mL
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC379805000

 View more versions of this product

Catalog No. AC379805000

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 229.7
Quantity 500mL
Density 1.0016g/mL
Appearance of Solution Clear colorless solution
CAS 7732-18-5
Health Hazard 2 GHS H Statement:
May intensify fire; oxidizer.
Risk of explosion if heated under confinement.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 2567-83-1
MDL Number MFCD00041977
Chemical Name or Material Tetraethylammonium perchlorate
Molecular Formula C8H20N·ClO4
Packaging Plastic bottle
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material Tetraethylammonium perchlorate
CAS 2567-83-1
CAS 7732-18-5
Molecular Formula C8H20N·ClO4
Formula Weight 229.7
MDL Number MFCD00041977
Accessories

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May intensify fire; oxidizer.
Risk of explosion if heated under confinement.

SDS & Safety Documents