Please login to your online account to display your discounted pricing

Tetraethylammonium p-toluenesulfonate, 98%, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  158795000

 View more versions of this product

Catalog No. AC158795000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystals or Crystalline Powder
Formula Weight 301.44
Quantity 500g
Color White to almost white
EINECS Number 211-992-1
TSCA TSCA
Melting Point 108° to 114°C
Chemical Name or Material Tetraethylammonium p-toluenesulfonate
Infrared Spectrum Authentic
Name Note 98%
Assay Percent Range 98%
Molecular Formula C15 H27 N O3 S
Linear Formula (C2H5)4N(O3SC6H4CH3)
MDL Number MFCD00011831
CAS 733-44-8
Packaging Plastic bottle
Health Hazard 1 HYGROSCOPIC

Specifications

Chemical Name or Material Tetraethylammonium p-toluenesulfonate
Name Note 98%
CAS 733-44-8
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C15 H27 N O3 S
Linear Formula (C2H5)4N(O3SC6H4CH3)
Formula Weight 301.44
MDL Number MFCD00011831
Accessories

HYGROSCOPIC

SDS & Safety Documents