Please login to your online account to display your discounted pricing

Tetraethylammonium p-toluenesulfonate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Tetraethylammonium p-toluenesulfonate
Name Note 98%
CAS 733-44-8
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C15 H27 N O3 S
Linear Formula (C2H5)4N(O3SC6H4CH3)
Formula Weight 301.44
MDL Number MFCD00011831
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC158795000

  SDS
thermo fisher scientific
158795000
Plastic bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystals or Crystalline Powder
Formula Weight 301.44
Quantity 500g
Color White to almost white
EINECS Number 211-992-1
TSCA TSCA
Melting Point 108° to 114°C
Chemical Name or Material Tetraethylammonium p-toluenesulfonate
Infrared Spectrum Authentic
Linear Formula (C2H5)4N(O3SC6H4CH3)
Name Note 98%
MDL Number MFCD00011831
Molecular Formula C15 H27 N O3 S
Assay Percent Range 98%
CAS 733-44-8
Packaging Plastic bottle
Health Hazard 1 HYGROSCOPIC

Phase transfer catalyst

Safety and Handling

HYGROSCOPIC