missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetraethylammonium Nitrate 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC420310100

 View more versions of this product

Catalog No. AC420310100

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.5%
Crystalline Powder
White
Tetraethylammonium nitrate
99%
1941-26-0
99%
192.26
10g
Glass bottle
C8H20N·NO3
(C2H5)4NNO3
MFCD00041978
Authentic

Chemical Identifiers

1941-26-0
C8H20N·NO3
192.26
Tetraethylammonium nitrate, 99%
99%
(C2H5)4NNO3
MFCD00041978
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
May intensify fire; oxidizer.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 217-725-5

TSCA : TSCA