Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraethylammonium nitrate, 35% w/w aq. soln.

Chemical Identifiers

Chemical Name or Material Tetraethylammonium nitrate
CAS 1941-26-0
Formula Weight 192.26
MDL Number MFCD00041978
UN Number UN3139
Beilstein 3918466
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41791AD

  SDS
alfa aesar™
41791AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA41791AK

  SDS
alfa aesar™
41791AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 192.26
Chemical Name or Material Tetraethylammonium nitrate
TSCA TSCA
Density 1.029g/mL
Physical Form Liquid
MDL Number MFCD00041978
CAS 1941-26-0
UN Number UN3139
Beilstein 3918466
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point >100°C
Concentration 35% w/w aq. soln.
Quantity 50mL
DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING LIQUID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.