Please login to your online account to display your discounted pricing

Tetraethylammonium iodide, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Tetraethylammonium iodide
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H20IN
Linear Formula (C2H5)4NI
Formula Weight 257.15
MDL Number MFCD00011829
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC216671000

  SDS
fisher scientific acros
216671000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 257.15
Quantity 100g
Color White to cream
Beilstein 04, 104
Fieser 12,465; 13,46
Merck Index 10, 9027
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C8H20IN
MDL Number MFCD00011829
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Chemical Name or Material Tetraethylammonium iodide
Linear Formula (C2H5)4NI
CAS 68-05-3
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Phase transfer catalyst

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.