Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraethylammonium hydroxide, 40% w/w Aqueous Solution

Chemical Identifiers

Chemical Name or Material Tetraethylammonium hydroxide
CAS 77-98-5
Formula Weight 147.26g/mol
UN Number UN3267
Merck Index 14,9201
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3081909

 
alfa aesar™
30819-09
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density 1.023g/mL
Refractive Index 1.4164
Boiling Point (dec.)
Melting Point 40° to 50°C
Odor Amine-like
Concentration 40% w/w aq. soln.
Quantity 10g
Chemical Name or Material Tetraethylammonium hydroxide
CAS 77-98-5
Physical Form Liquid
Formula Weight 147.26g/mol
UN Number UN3267
Merck Index 14,9201
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.