missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetraethylammonium Hydroxide (35% in Water), TCI America™

Manufacturer:  TCI America T2589500ML

 View more versions of this product

Catalog No. T2589500ML



Specifications

Specifications

Tetraethylammonium Hydroxide (35% in Water)
500mL
C8H21NO
3267
LRGJRHZIDJQFCL-UHFFFAOYSA-M
tetraethylazanium;hydroxide
6509
Clear Liquid at 20°C
Yellow
77-98-5
MFCD00009024
tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium
CC[N+](CC)(CC)CC.[OH-]
147.262
147.26

Chemical Identifiers

77-98-5
147.262
LRGJRHZIDJQFCL-UHFFFAOYSA-M
6509
CC[N+](CC)(CC)CC.[OH-]
C8H21NO
MFCD00009024
tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium
tetraethylazanium;hydroxide
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0186

RTECSNumber : KH3150000

TSCA : Yes

Documents