Please login to your online account to display your discounted pricing

Tetraethylammonium hydroxide, 25% in water, ACROS Organics™


Quantity 25g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  420290250

 View more versions of this product

Catalog No. AC420290250

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Solution
Formula Weight 147.26
Quantity 25g
Color Clear colorless to light yellow
EINECS Number 201-073-3
TSCA TSCA
Assay 24 to 26%
Density 1.0200g/mL
RTECS Number KH3150000
Chemical Name or Material Tetraethylammonium hydroxide
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
CAS 7732-18-5
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Plastic bottle
Name Note 25% in Water
Molecular Formula C8H21NO
Linear Formula (C2H5)4NOH
MDL Number MFCD00009024
CAS 77-98-5

Specifications

Chemical Name or Material Tetraethylammonium hydroxide
Name Note 25% in Water
CAS 77-98-5
CAS 7732-18-5
Assay 24 to 26%
Molecular Formula C8H21NO
Linear Formula (C2H5)4NOH
Formula Weight 147.26
MDL Number MFCD00009024
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.

SDS & Safety Documents