missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraethylammonium hydroxide, 25% w/w in methanol

Manufacturer:  Alfa Aesar™ 1632830

 View more versions of this product

Catalog No. AA1632830



Specifications

Specifications

Tetraethylammonium hydroxide
16° (60°F)
1.380
250g
C8H21NO
UN2924
14,9201
25% w/w in methanol
CC[N+](CC)(CC)CC.[OH-]
147.262
147.26
0.88g/mL
Like alcohol
Air Sensitive
77-98-5
MFCD00009024
3912961
tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium
LRGJRHZIDJQFCL-UHFFFAOYSA-M
tetraethylazanium;hydroxide
6509
Liquid

Chemical Identifiers

77-98-5
147.262
LRGJRHZIDJQFCL-UHFFFAOYSA-M
6509
CC[N+](CC)(CC)CC.[OH-]
C8H21NO
MFCD00009024
tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium
tetraethylazanium;hydroxide
Safety and Handling

Safety and Handling

GHS H Statement
H224-H300-H310-H330-H370-H314-H318
Extremely flammable liquid and vapor.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes damage to organs.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see ... on this label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

TSCA : TSCA

SDS Documents