missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetraethylammonium Hydroxide, 10% in Water, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 420285000

 View more versions of this product

Catalog No. AC420285000


 

Specifications Description & Specifications

Specifications

Tetraethylammonium hydroxide
77-98-5,7732-18-5
9.5 to 10.5%
(C2H5)4NOH
1.0200g/mL
15, 9346
Clear colorless
10% in Water
Liquid
C8H21NO
147.26
MFCD00009024
Plastic bottle
500g

Chemical Identifiers

77-98-5,7732-18-5
(C2H5)4NOH
MFCD00009024
9.5 to 10.5%
C8H21NO
147.26
Tetraethylammonium hydroxide, 10% in Water

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-073-3

RTECSNumber : KH3150000

TSCA : TSCA