Please login to your online account to display your discounted pricing

Tetraethylammonium hydrogensulfate, 99+%, HPLC grade, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC343820500

 View more versions of this product

Catalog No. AC343820500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 227.32
Purity Grade HPLC Grade
Physical Form Powder
Assay ≥99.0% (on dry substance)
Quantity 50g
Color White to off-white
Absorbance 0.02 max. (0.4 M in HPLC water) at 250nm
Chemical Name or Material Tetraethylammonium hydrogensulfate
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 16873-13-5
MDL Number MFCD00036150
Molecular Formula C8H20N·HSO4
Assay Percent Range 99+%
Packaging Glass bottle
Infrared Spectrum Authentic
Water 0.5% max. (K.F.)

Specifications

Chemical Name or Material Tetraethylammonium hydrogensulfate
CAS 16873-13-5
Purity Grade HPLC Grade
Assay ≥99.0% (on dry substance)
Assay Percent Range 99+%
Molecular Formula C8H20N·HSO4
Formula Weight 227.32
MDL Number MFCD00036150
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents