Learn More
Tetraethylammonium hydrogensulfate, 99+%, HPLC grade, Thermo Scientific™
Chemical Identifiers
CAS | 16873-13-5 |
---|---|
Molecular Formula | C8H21NO4S |
Molecular Weight (g/mol) | 227.32 |
MDL Number | MFCD00036150 |
InChI Key | CREVBWLEPKAZBH-UHFFFAOYSA-M |
Synonym | tetraethylammonium hydrogensulfate, tetraethylammonium hydrogen sulphate, tetraethylammonium hydrogen sulfate, n,n,n-triethylethanaminium hydrogen sulfate 1:1:1, hydrogen sulfate; tetraethylazanium, tertraethylammonium hydrogen sulfate, tetraethyl ammonium hydrogen sulphate |
PubChem CID | 167582 |
IUPAC Name | hydrogen sulfate;tetraethylazanium |
SMILES | OS([O-])(=O)=O.CC[N+](CC)(CC)CC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC343820050
|
Thermo Scientific Chemicals
343820050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
for ion pair chromatography, Phase transfer catalystChemical Identifiers
16873-13-5 | |
227.32 | |
CREVBWLEPKAZBH-UHFFFAOYSA-M | |
167582 | |
OS([O-])(=O)=O.CC[N+](CC)(CC)CC |
C8H21NO4S | |
MFCD00036150 | |
tetraethylammonium hydrogensulfate, tetraethylammonium hydrogen sulphate, tetraethylammonium hydrogen sulfate, n,n,n-triethylethanaminium hydrogen sulfate 1:1:1, hydrogen sulfate; tetraethylazanium, tertraethylammonium hydrogen sulfate, tetraethyl ammonium hydrogen sulphate | |
hydrogen sulfate;tetraethylazanium |
Specifications
16873-13-5 | |
Authentic | |
Glass bottle | |
MFCD00036150 | |
tetraethylammonium hydrogensulfate, tetraethylammonium hydrogen sulphate, tetraethylammonium hydrogen sulfate, n,n,n-triethylethanaminium hydrogen sulfate 1:1:1, hydrogen sulfate; tetraethylazanium, tertraethylammonium hydrogen sulfate, tetraethyl ammonium hydrogen sulphate | |
OS([O-])(=O)=O.CC[N+](CC)(CC)CC | |
227.32 | |
227.32 | |
HPLC |
0.02 max. (0.4 M in HPLC water) at 250nm, 0.04 max. (0.4 M in HPLC water) at 230nm, 0.04 max. (0.4 M in HPLC water) at 240nm, 0.05 max. (0.4 M in HPLC water) at 220nm, 0.1 max. (0.4 M in HPLC water) at 210nm | |
99+% | |
C8H21NO4S | |
5g | |
CREVBWLEPKAZBH-UHFFFAOYSA-M | |
hydrogen sulfate;tetraethylazanium | |
167582 | |
99+% | |
Tetraethylammonium hydrogensulfate |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 240-899-9
RUO â Research Use Only