missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetraethylammonium hydrogensulfate, 99+%, HPLC grade, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC343820050

 View more versions of this product

Catalog No. AC343820050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Tetraethylammonium hydrogensulfate
5g
16873-13-5
Glass bottle
Powder
≥99.0% (on dry substance)
C8H20N·HSO4
MFCD00036150
White to off-white
0.02 max. (0.4 M in HPLC water) at 250nm
Authentic
HPLC Grade
0.5% max. (K.F.)
99+%
227.32

Chemical Identifiers

16873-13-5
C8H20N·HSO4
MFCD00036150
≥99.0% (on dry substance)
99+%
227.32
Tetraethylammonium hydrogensulfate
HPLC Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.