missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraethylammonium hexafluorophosphate, 98%

Manufacturer:  Alfa Aesar™ 1625609

 View more versions of this product

Catalog No. AA1625609



Specifications

Specifications

10g
Hygroscopic
C8H20F6NP
UN2811
KLKUOIXSIDDDCN-UHFFFAOYSA-N
tetraethylazanium;hexafluorophosphate
9885
Crystalline
Tetraethylammonium hexafluorophosphate
429-07-2
MFCD00043179
tetraethylammonium hexafluorophosphate for electrochemical analysis, ethanaminium nnn-triethyl- hexafluorophosphate 1-9ci, ethanaminium nnn-triethyl- hexafluorophosphate 1-, ethanaminiumnn-triethyl- hexafluorophosphate 1-, nnn-triethylethanaminium hexafluorophosphate, ammonium tetraethyl- hexafluorophosphate 1-, tetraethylammonium hexafluorophosphate, tetraethylammonium hexafluorophosphate v, tetraethylazanium hexafluorophosphate
CC[N+](CC)(CC)CC.F[P-](F)(F)(F)(F)F
275.219
275.23
98%

Chemical Identifiers

429-07-2
275.219
KLKUOIXSIDDDCN-UHFFFAOYSA-N
9885
CC[N+](CC)(CC)CC.F[P-](F)(F)(F)(F)F
C8H20F6NP
MFCD00043179
tetraethylammonium hexafluorophosphate for electrochemical analysis, ethanaminium nnn-triethyl- hexafluorophosphate 1-9ci, ethanaminium nnn-triethyl- hexafluorophosphate 1-, ethanaminiumnn-triethyl- hexafluorophosphate 1-, nnn-triethylethanaminium hexafluorophosphate, ammonium tetraethyl- hexafluorophosphate 1-, tetraethylammonium hexafluorophosphate, tetraethylammonium hexafluorophosphate v, tetraethylazanium hexafluorophosphate
tetraethylazanium;hexafluorophosphate
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

SDS Documents