missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetraethylammonium fluoride hydrate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 216660250

 View more versions of this product

Catalog No. AC216660250


 

Specifications Description & Specifications

Specifications

≥97.5% (on dry substance)
149.14
White to beige
C8H20FN·xH2O
(C2H5)4NF·xH2O
Tetraethylammonium fluoride hydrate
98%
28% max. (K.F.)
Adhering Crystals or Crystals or Crystalline Solid
25g
Authentic
98%
MFCD00149990
98330-04-2
Glass bottle

Chemical Identifiers

98330-04-2
C8H20FN·xH2O
149.14
Tetraethylammonium fluoride hydrate, 98%
98%
(C2H5)4NF·xH2O
MFCD00149990
≥97.5% (on dry substance)

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning