missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraethylammonium chloride monohydrate, 98+%

Phase transfer catalyst

Manufacturer:  Alfa Aesar™ 3081818

 View more versions of this product

Catalog No. AA3081818



Specifications

Specifications

Tetraethylammonium chloride monohydrate
1.0801g/mL
C8H22ClNO
3563247
CALLTGJPWMIDPC-UHFFFAOYSA-M
tetraethylazanium;chloride;hydrate
16211205
Crystalline
50g
68696-18-4
≥98%
MFCD00149992
14,9200
CC[N+](CC)(CC)CC.O.[Cl-]
183.72
183.72 (165.71 Anhydrous)
~110°C

Chemical Identifiers

68696-18-4
183.72
CALLTGJPWMIDPC-UHFFFAOYSA-M
tetraethylazanium;chloride;hydrate
C8H22ClNO
MFCD00149992
16211205
CC[N+](CC)(CC)CC.O.[Cl-]
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : BS6125000

TSCA : TSCA

SDS Documents