missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetraethylammonium chloride hydrate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1521522

 View more versions of this product

Catalog No. AAA1521522



Description

Description

Tetraethylammonium chloride monohydrate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraethylammonium chloride monohydrate is used as pharmaceutical intermediate.

Solubility
Soluble in water (1410 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

Tetraethylammonium chloride monohydrate
∼110°C
C8H22ClNO
100 g
14,9200
Soluble in water (1410g/L at 20°C).
CC[N+](CC)(CC)CC.O.[Cl-]
183.72
183.72 (165.71 Anhydrous)
68696-18-4
1.0801
MFCD00149992
3563247
tetraethylammonium chloride hydrate, tetraethylammonium chloride monohydrate, tetraethylazanium chloride hydrate, tetraethylammonium hydrate chloride, c8h20n.cl.h2o, tetraethylammoniumchloride hydrate, tetraethyl ammonium chloride hydrate, tetraethylammonium chloride hydrate, cryst, n,n,n-triethylethanaminium chloride-water 1/1/1, tetraethylammonium chloride, monohydrate
CALLTGJPWMIDPC-UHFFFAOYSA-M
tetraethylazanium;chloride;hydrate
16211205
98%

Chemical Identifiers

68696-18-4
183.72
CALLTGJPWMIDPC-UHFFFAOYSA-M
16211205
CC[N+](CC)(CC)CC.O.[Cl-]
C8H22ClNO
MFCD00149992
tetraethylammonium chloride hydrate, tetraethylammonium chloride monohydrate, tetraethylazanium chloride hydrate, tetraethylammonium hydrate chloride, c8h20n.cl.h2o, tetraethylammoniumchloride hydrate, tetraethyl ammonium chloride hydrate, tetraethylammonium chloride hydrate, cryst, n,n,n-triethylethanaminium chloride-water 1/1/1, tetraethylammonium chloride, monohydrate
tetraethylazanium;chloride;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 200-267-5

RTECSNumber : BS6125000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only