Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraethylammonium chloride monohydrate, 98+%

Phase transfer catalyst

$244.74 - $244.74

Chemical Identifiers

Chemical Name or Material Tetraethylammonium chloride monohydrate
CAS 68696-18-4
Formula Weight 183.72 (165.71 Anhydrous)
MDL Number MFCD00149992
Beilstein 3563247
Merck Index 14,9200
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3081818

 
alfa aesar™
3081818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3081830

 
alfa aesar™
3081830
250g Each for $244.74
Description & Specifications

Specifications

Formula Weight 183.72 (165.71 Anhydrous)
Chemical Name or Material Tetraethylammonium chloride monohydrate
TSCA TSCA
Density 1.0801g/mL
Physical Form Crystalline
Assay ≥98%
MDL Number MFCD00149992
CAS 68696-18-4
Beilstein 3563247
Merck Index 14,9200
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈110°C
Quantity 50g
RTECS Number BS6125000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.