Learn More
Tetraethylammonium Chloride Hydrate 99%, ACROS Organics™
Manufacturer: Acros Organics 150905000
Description
Phase transfer catalystSpecifications
Tetraethylammonium chloride hydrate | |
10% max. | |
White | |
99% | |
99% | |
(C2H5)4NCl·xH2O | |
15, 9345 | |
Solubility in water: in water: 1410g/L (25°C). Other solubilities: freely soluble in alcohol, chloroform and acetone, slightly soluble in benzene and ether | |
CC[N+](CC)(CC)CC.[Cl-] | |
165.705 | |
CHEBI:78161 | |
Crystalline Powder |
Authentic | |
Glass bottle | |
500g | |
56-34-8 | |
C8H20ClN | |
MFCD00149992 | |
tetraethylammonium chloride, tetraethyl ammonium chloride, etamon chloride, tea chloride, teac, n,n,n-triethylethanaminium chloride, tetraethylammoniumchloride, ethanaminium, n,n,n-triethyl-, chloride, tetraethyl-ammonium chloride, unii-8b82z83xnn | |
YMBCJWGVCUEGHA-UHFFFAOYSA-M | |
tetraethylazanium;chloride | |
5946 | |
165.71 | |
99% |
Chemical Identifiers
56-34-8 | |
165.705 | |
YMBCJWGVCUEGHA-UHFFFAOYSA-M | |
5946 | |
tetraethylazanium;chloride |
C8H20ClN | |
MFCD00149992 | |
tetraethylammonium chloride, tetraethyl ammonium chloride, etamon chloride, tea chloride, teac, n,n,n-triethylethanaminium chloride, tetraethylammoniumchloride, ethanaminium, n,n,n-triethyl-, chloride, tetraethyl-ammonium chloride, unii-8b82z83xnn | |
CHEBI:78161 | |
CC[N+](CC)(CC)CC.[Cl-] |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
EINECSNumber : 200-267-5
RTECSNumber : BS6125000
TSCA : TSCA