missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetraethylammonium bromide, 98%, Thermo Scientific Chemicals

Phase transfer catalyst

Supplier:  Thermo Scientific Chemicals 161072500

 View more versions of this product

Catalog No. AC161072500



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Specifications

Tetraethylammonium bromide
71-91-0
97.5% min. (Argentometry)
C8H20BrN
MFCD00011825
04,104
15,9344
Solubility in water: 2795g/L (25°C). Other solubilities: freely soluble in alcohol,chloroform and acetone,slightly soluble in benzene
CC[N+](CC)(CC)CC.[Br-]
210.16
210.16
98%
285.0°C
Plastic bottle
(C2H5)4NBr
250 g
06,568; 13,46
tetraethylammonium bromide, teab, tea bromide, tetrylammonium bromide, sympatektoman, tetranium, etylon, bromethyl, beparon, etambro
HWCKGOZZJDHMNC-UHFFFAOYSA-M
tetraethylazanium;bromide
6285
98%

Chemical Identifiers

71-91-0
210.16
HWCKGOZZJDHMNC-UHFFFAOYSA-M
6285
CC[N+](CC)(CC)CC.[Br-]
C8H20BrN
MFCD00011825
tetraethylammonium bromide, teab, tea bromide, tetrylammonium bromide, sympatektoman, tetranium, etylon, bromethyl, beparon, etambro
tetraethylazanium;bromide
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 200-769-4

RTECSNumber : BS5950000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only