Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraethylammonium borohydride

Chemical Identifiers

Chemical Name or Material Tetraethylammonium borohydride
CAS 17083-85-1
Linear Formula (C2H5)4NBH4
Formula Weight 145.1
MDL Number MFCD00031600
UN Number UN3131
Beilstein 3693695
Synonym Tetraethylammonium tetrahydridoborate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3257304

  SDS
alfa aesar™
3257304
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 145.1
MDL Number MFCD00031600
Density 0.926g/mL
UN Number UN3131
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P370+P378i-P402+P404
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
In case of fire: Use for extinction: Fire-extinguishing powder.
Store in a dry place.
Store in a closed container.
Chemical Name or Material Tetraethylammonium borohydride
Health Hazard 2 GHS H Statement
H261-H314-H318-H302
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 1 Danger
Melting Point ≈225°C (dec.)
Odor Odorless
Physical Form Powder
RTECS Number BS5850000
Beilstein 3693695
Sensitivity Moisture Sensitive
Color White
Synonym Tetraethylammonium tetrahydridoborate
DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE SOLID, CORROSIVE, N.O.S.
TSCA TSCA
CAS 17083-85-1
Quantity 2g
Linear Formula (C2H5)4NBH4
Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P370+P378i-P402+P404
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
In case of fire: Use for extinction: Fire-extinguishing powder.
Store in a dry place.
Store in a closed container.

GHS H Statement
H261-H314-H318-H302
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.