missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetraethylammonium acetate tetrahydrate, 97+%, Thermo Scientific™
Phase transfer catalyst
Supplier: Thermo Scientific Chemicals 194945000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Phase transfer catalystSpecifications
Tetraethylammonium acetate tetrahydrate | |
60°C | |
97+% | |
C10H23NO2·4H2O | |
500g | |
Solubility in water: soluble. | |
CC[N+](CC)(CC)CC.CC(=O)[O-].O.O.O.O | |
261.35 | |
261.35 |
67533-12-4 | |
Authentic | |
Glass bottle | |
(C2H5)4N(O2CCH3)·4H2O | |
tetraethylammonium acetate tetrahydrate, ethanaminium, n,n,n-triethyl-, acetate, tetrahydrate, tetraethylazanium acetate tetrahydrate, tetraethylazanium ethanoate tetrahydrate, acetate ion tetraethylammonium tetrahydrate, ethanaminium,n,n,n-triethyl-,acetate,tetrahydrate | |
QNOAKQGNSWGYOE-UHFFFAOYSA-M | |
tetraethylazanium;acetate;tetrahydrate | |
2724098 | |
97+% |
Safety and Handling
GHS Signal Word: Danger
RUO â Research Use Only