missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetraethoxysilane, 99+%, Alfa Aesar™

As a binding agent and as a polymerization catalyst

Manufacturer:  Alfa Aesar 1408257

 View more versions of this product

Catalog No. AA1408257



Specifications

Specifications

Tetraethoxysilane
C8H20O4Si
MFCD00009062
1422225
tetraethyl orthosilicate, tetraethoxysilane, teos, ethyl silicate, silicon ethoxide, silicon tetraethoxide, silane, tetraethoxy, dynasil a, tetraethoxysilicon, ethyl orthosilicate
CCO[Si](OCC)(OCC)OCC
208.329
208.33
≥99%
Pleasant
Moisture Sensitive
Colorless
4 x 500g
78-10-4
(C2H5O)4Si
UN1292
14,3851
BOTDANWDWHJENH-UHFFFAOYSA-N
tetraethyl silicate
6517
Liquid
0.934
1.3820
168°C
-85°C
46°C (116°F)

Chemical Identifiers

78-10-4
208.329
BOTDANWDWHJENH-UHFFFAOYSA-N
6517
CCO[Si](OCC)(OCC)OCC
C8H20O4Si
MFCD00009062
tetraethyl orthosilicate, tetraethoxysilane, teos, ethyl silicate, silicon ethoxide, silicon tetraethoxide, silane, tetraethoxy, dynasil a, tetraethoxysilicon, ethyl orthosilicate
tetraethyl silicate
Safety and Handling

Safety and Handling

GHS H Statement
H226-H332-H319-H335;
Flammable liquid and vapor.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TETRAETHYL SILICATE

RTECSNumber : VV9450000

TSCA : TSCA

Documents