Please login to your online account to display your discounted pricing

Tetradecylbenzene 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  124680050

 View more versions of this product

Catalog No. AC124680050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 274.49
Physical Form Liquid
Quantity 5g
Color Clear colorless to yellow
TSCA TSCA
EINECS Number 215-950-3
Density 0.8500g/mL
Refractive Index 1.4805 to 1.4825 (20°C, 589nm)
CAS 1459-10-5
Linear Formula C6H5(CH2)13CH3
Name Note 99%
Chemical Name or Material Tetradecylbenzene
Assay Percent Range 98%
MDL Number MFCD00015080
Packaging Glass bottle
Synonym 1-Phenyltetradecane
Molecular Formula C20H34
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Tetradecylbenzene
Name Note 99%
CAS 1459-10-5
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C20H34
Linear Formula C6H5(CH2)13CH3
Formula Weight 274.49
MDL Number MFCD00015080
Synonym 1-Phenyltetradecane