Please login to your online account to display your discounted pricing

Tetradecylbenzene 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Tetradecylbenzene
Name Note 99%
CAS 1459-10-5
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C20H34
Linear Formula C6H5(CH2)13CH3
Formula Weight 274.49
MDL Number MFCD00015080
Synonym 1-Phenyltetradecane
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC124680250

 
thermo fisher scientific
124680250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC124680050

 
thermo fisher scientific
124680050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 274.49
Quantity 25g
Color Clear colorless to yellow
EINECS Number 215-950-3
TSCA TSCA
Density 0.8500g/mL
Chemical Name or Material Tetradecylbenzene
Packaging Glass bottle
Refractive Index 1.4805 to 1.4825 (20°C, 589nm)
Name Note 99%
Assay Percent Range 98%
Molecular Formula C20H34
Linear Formula C6H5(CH2)13CH3
MDL Number MFCD00015080
CAS 1459-10-5
Synonym 1-Phenyltetradecane
Infrared Spectrum Authentic