Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetradecanonitrile, 98%

$55.55 - $706.53

Chemical Identifiers

Chemical Name or Material Tetradecanonitrile
CAS 629-63-0
Molecular Formula CH3(CH2)12CN
Formula Weight 209.38
MDL Number MFCD00019848
Beilstein 1705598
Synonym Myristonitrile; Myrystyl nitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1970414

 
alfa aesar™
A1970414
25g Each for $55.55

AAA1970422

 
alfa aesar™
A1970422
100g Each for $180.09

AAA1970436

 
alfa aesar™
A1970436
500g Each for $706.53
Description & Specifications

Specifications

Formula Weight 209.38
Chemical Name or Material Tetradecanonitrile
TSCA TSCA
Assay 98%
Molecular Formula CH3(CH2)12CN
MDL Number MFCD00019848
CAS 629-63-0
Beilstein 1705598
Synonym Myristonitrile; Myrystyl nitrile
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 226° to 227°C (100mmHg)
Melting Point ≈19°C
Odor Odorless
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.