Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetracyclohexyltin

Chemical Identifiers

Chemical Name or Material Tetracyclohexyltin
CAS 1449-55-4
Formula Weight 451.31
MDL Number MFCD00014278
UN Number UN3146
Synonym Tetracyclohexylstannane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1882203

  SDS
alfa aesar™
1882203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1882206

  SDS
alfa aesar™
1882206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1882214

  SDS
alfa aesar™
1882214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 451.31
Chemical Name or Material Tetracyclohexyltin
Melting Point 241° to 250°C
Physical Form Powder
MDL Number MFCD00014278
CAS 1449-55-4
Quantity 1g
UN Number UN3146
Synonym Tetracyclohexylstannane
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.