missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetracyclohexyltin

Manufacturer:  Alfa Aesar™ 1882214

 View more versions of this product

Catalog No. AA1882214



Specifications

Specifications

Tetracyclohexyltin
1449-55-4
C24H44Sn
UN3146
JUISPCSEIXBMNI-UHFFFAOYSA-N
tetracyclohexylstannane
74056
Powder
241°C to 250°C
25g
MFCD00014278
tetracyclohexyltin, stannane, tetracyclohexyl, tetracyclohexylstananne, tetracyclohexyl tin, tin, tetracyclohexyl, acmc-1c0rn
C1CCC(CC1)[Sn](C2CCCCC2)(C3CCCCC3)C4CCCCC4
451.326
451.31

Chemical Identifiers

1449-55-4
451.326
JUISPCSEIXBMNI-UHFFFAOYSA-N
74056
C1CCC(CC1)[Sn](C2CCCCC2)(C3CCCCC3)C4CCCCC4
C24H44Sn
MFCD00014278
tetracyclohexyltin, stannane, tetracyclohexyl, tetracyclohexylstananne, tetracyclohexyl tin, tin, tetracyclohexyl, acmc-1c0rn
tetracyclohexylstannane
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.

SDS Documents