Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetracycline hydrochloride, 10mg/mL in 100% methanol, sterile-filtered

Chemical Identifiers

Chemical Name or Material Tetracycline hydrochloride
CAS 64-75-5
Name Note 10mg/mL in 100% methanol, sterile-filtered
MDL Number MFCD00078142
UN Number UN1230
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67277AD

  SDS
alfa aesar™
J67277AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67277AE

  SDS
alfa aesar™
J67277AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 64-75-5
MDL Number MFCD00078142
Health Hazard 1 Danger
RTECS Number PC1400000
Physical Form Liquid
Health Hazard 2 GHS H Statement
H225-H311-H331-H370-H361
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Suspected of damaging fertility or the unborn child.
TSCA TSCA
Quantity 50mL
Name Note 10mg/mL in 100% methanol, sterile-filtered
Storage Note 1 -30° to -10°C
UN Number UN1230
DOT Information Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: METHANOL
Chemical Name or Material Tetracycline hydrochloride
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H311-H331-H370-H361
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Suspected of damaging fertility or the unborn child.

-30° to -10°C