Learn More
Tetracyanoethylene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 138050250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Oxidation reagentSpecifications
Tetracyanoethylene | |
196.0°C to 201.0°C | |
Authentic | |
Glass bottle | |
(NC)2C=C(CN)2 | |
01,1133; 02,397; 05,647; 06,567; 13,289 | |
tetracyanoethylene, tetracyanoethene, ethylenetetracarbonitrile, tcne, ethenetetracarbonitrile, ethene, tetracyano, 1,1,2,2-ethenetetracarbonitrile, tetrakyanethylen, 1,1,2,2-tetracyanoethene, 1,1,2,2-tetracyanoethylene | |
NLDYACGHTUPAQU-UHFFFAOYSA-N | |
ethene-1,1,2,2-tetracarbonitrile | |
12635 | |
98% |
670-54-2 | |
223.0°C | |
97.5% min. (Ex N) | |
C6N4 | |
25 g | |
15,934 | |
Solubility in water: hydrolyzes (forming hcn) in water. Other solubilities: slightly soluble in: ether, benzene,, carbon tetrachloride, soluble in: acetone | |
C(#N)C(=C(C#N)C#N)C#N | |
128.09 | |
128.09 | |
Crystalline Powder, Crystals or Chunks |
Chemical Identifiers
670-54-2 | |
128.09 | |
tetracyanoethylene, tetracyanoethene, ethylenetetracarbonitrile, tcne, ethenetetracarbonitrile, ethene, tetracyano, 1,1,2,2-ethenetetracarbonitrile, tetrakyanethylen, 1,1,2,2-tetracyanoethene, 1,1,2,2-tetracyanoethylene | |
ethene-1,1,2,2-tetracarbonitrile |
C6N4 | |
NLDYACGHTUPAQU-UHFFFAOYSA-N | |
12635 | |
C(#N)C(=C(C#N)C#N)C#N |
Safety and Handling
GHS H Statement
Fatal in contact with skin.
Fatal if swallowed.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Rinse mouth.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable
GHS Signal Word: Danger
EINECSNumber : 211-578-
RUO – Research Use Only