missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetracosanol, 99+%, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific AC258151000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Tetracosanol | |
White | |
218°C | |
Authentic | |
CH3(CH2)23OH | |
Glass bottle | |
100mg | |
TYWMIZZBOVGFOV-UHFFFAOYSA-N | |
tetracosan-1-ol | |
10472 | |
354.65 | |
99+% |
506-51-4 | |
75°C to 77°C | |
99+% | |
C24H50O | |
MFCD00042662 | |
1-tetracosanol, 2n0pi37ioc, lignoceric alcohol, lignocerol, lignoceryl alcohol, n-tetracosanol, n-tetracosanol-1, tetracosanol, tetracosyl alcohol, unii-2n0pi37ioc | |
Solubility in water: .. Other solubilities: soluble in hydrocarbons,alcohols,ether,ketones,,specially when hot | |
CCCCCCCCCCCCCCCCCCCCCCCCO | |
354.66 | |
CHEBI:77413 | |
Shiny Flakes or Fluffy Powder |
Chemical Identifiers
506-51-4 | |
354.66 | |
TYWMIZZBOVGFOV-UHFFFAOYSA-N | |
10472 | |
tetracosan-1-ol |
C24H50O | |
MFCD00042662 | |
1-tetracosanol, 2n0pi37ioc, lignoceric alcohol, lignocerol, lignoceryl alcohol, n-tetracosanol, n-tetracosanol-1, tetracosanol, tetracosyl alcohol, unii-2n0pi37ioc | |
CHEBI:77413 | |
CCCCCCCCCCCCCCCCCCCCCCCCO |
Safety and Handling
EINECSNumber : 208-043-9
RUO – Research Use Only