missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetracontane, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 309165000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tetracontane | |
80.0°C to 83.0°C | |
Authentic | |
Glass bottle | |
CH3(CH2)38CH3 | |
500 mg | |
n-tetracontane, unii-i79s9ixb9y, alkane c40, i79s9ixb9y, tetracontane, analytical standard, acmc-209jlw, n-tetracontane 500mg, tetracontane gc | |
KUPLEGDPSCCPJI-UHFFFAOYSA-N | |
tetracontane | |
20149 | |
98% |
4181-95-7 | |
White | |
97.5% min. (GC) | |
C40H82 | |
MFCD00015267 | |
01,III,592 | |
Solubility in water: insoluble. Other solubilities: soluble in petroleum ether | |
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC | |
563.10 | |
563.1 | |
Powder or Flaky Solid |
Chemical Identifiers
4181-95-7 | |
563.10 | |
KUPLEGDPSCCPJI-UHFFFAOYSA-N | |
20149 | |
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC |
C40H82 | |
MFCD00015267 | |
n-tetracontane, unii-i79s9ixb9y, alkane c40, i79s9ixb9y, tetracontane, analytical standard, acmc-209jlw, n-tetracontane 500mg, tetracontane gc | |
tetracontane |
Safety and Handling
EINECSNumber : 224-055-7
RUO – Research Use Only