Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetrachlorothiophene, 98%

Chemical Identifiers

Chemical Name or Material Tetrachlorothiophene
CAS 6012-97-1
Molecular Formula C4Cl4S
Formula Weight 221.92
MDL Number MFCD00005422
UN Number UN2811
Beilstein 118374
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0312703

 
alfa aesar™
L0312703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0312706

 
alfa aesar™
L0312706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0312714

 
alfa aesar™
L0312714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 221.92
Molecular Formula C4Cl4S
Chemical Name or Material Tetrachlorothiophene
MDL Number MFCD00005422
CAS 6012-97-1
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Assay 98%
UN Number UN2811
Beilstein 118374
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Health Hazard 3 GHS P Statement
P280h-P273-P270-P309-P310-P501a
Wear protective gloves/clothing.
Avoid release to the environment.
Do not eat, drink or smoke when using this product.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 232° to 233°C
Melting Point 26° to 30°C
Flash Point >110°C (230°F)
Odor Characteristic
Quantity 1g
RTECS Number XN0350000
Safety and Handling

Danger

GHS P Statement
P280h-P273-P270-P309-P310-P501a
Wear protective gloves/clothing.
Avoid release to the environment.
Do not eat, drink or smoke when using this product.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.