Please login to your online account to display your discounted pricing

Tetrachlorophthalic Anhydride 98%, ACROS Organics™


Quantity 2.5kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC157790025

 View more versions of this product

Catalog No. AC157790025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 285.89
Physical Form Flakes
Melting Point >254.5°C
Quantity 2.5kg
Color White to off-white
Beilstein 17, 484
Fieser 15,300
TSCA TSCA
EINECS Number 204-171-4
RTECS Number TI3450000
Chemical Name or Material Tetrachlorophthalic anhydride
Name Note 98%
Infrared Spectrum Authentic
Assay Percent Range 98%
Iron (Fe) 10ppm max.
Molecular Formula C8Cl4O3
MDL Number MFCD00005920
CAS 117-08-8
Synonym 4,5,6,7-Tetrachloro-1,3-isobenzofurandione
Packaging Plastic bottle
Additional Information alkali insolubles (10g in NaOH): < 0.5%
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause cancer.
CAS 118-74-1
Health Hazard 3 GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Tetrachlorophthalic anhydride
Name Note 98%
CAS 117-08-8
CAS 118-74-1
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8Cl4O3
Formula Weight 285.89
MDL Number MFCD00005920
Synonym 4,5,6,7-Tetrachloro-1,3-isobenzofurandione
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause cancer.

SDS & Safety Documents