Please login to your online account to display your discounted pricing

Tetrachlorophthalic Anhydride 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Tetrachlorophthalic anhydride
Name Note 98%
CAS 117-08-8
CAS 118-74-1
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8Cl4O3
Formula Weight 285.89
MDL Number MFCD00005920
Synonym 4,5,6,7-Tetrachloro-1,3-isobenzofurandione
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC157790025

  SDS
thermo fisher scientific
AC157790025
Plastic bottle 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC157790010

  SDS
fisher scientific acros
157790010
Plastic bottle 1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC157792500

  SDS
fisher scientific acros
157792500
Plastic bottle 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 285.89
Melting Point >254.5°C
Physical Form Flakes
Quantity 2.5kg
Color White to off-white
Beilstein 17, 484
Fieser 15,300
TSCA TSCA
RTECS Number TI3450000
EINECS Number 204-171-4
Synonym 4,5,6,7-Tetrachloro-1,3-isobenzofurandione
Additional Information alkali insolubles (10g in NaOH): < 0.5%
CAS 118-74-1
Health Hazard 2 GHS H Statement:
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause cancer.
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Packaging Plastic bottle
Name Note 98%
CAS 117-08-8
Chemical Name or Material Tetrachlorophthalic anhydride
Assay Percent Range 98%
Molecular Formula C8Cl4O3
MDL Number MFCD00005920
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause cancer.