missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrachloroethylene, Ultrapure, Spectrophotometric Grade, >99%, Alfa Aesar™

Manufacturer:  Alfa Aesar 32437K2

 View more versions of this product

Catalog No. AA32437K2



Specifications

Specifications

Tetrachloroethylene, Ultrapure
1L
>99%
MFCD00000834
CYTYCFOTNPOANT-UHFFFAOYSA-N
1,1,2,2-tetrachloroethene
31373
165.83
Sprectrophotometric
1.623 g/mL
127-18-4
C2Cl4
tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
C(=C(Cl)Cl)(Cl)Cl
165.822
CHEBI:17300
Liquid

Chemical Identifiers

127-18-4
165.822
CYTYCFOTNPOANT-UHFFFAOYSA-N
31373
1,1,2,2-tetrachloroethene
C2Cl4
MFCD00000834
tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
CHEBI:17300
C(=C(Cl)Cl)(Cl)Cl
Safety and Handling

Safety and Handling

EINECSNumber : 204-825-9

SDS Documents