Learn More
Tetrachloroethylene, Ultrapure, Spectrophotometric Grade, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 032437M1
Description
Tetrachloroethylene is used as solvent. It is involved in dry cleaning as well as in the degreasing of metal parts. It finds application as a neutrino detector. It is utilized as a paint stripper and a spot remover in consumer products. It acts as a precursor for the manufacture of other chemicals. It plays an important role in water repellants, paint removers, printing inks, glues, sealants, polishes and lubricants. As a film-forming electrolyte additive, it is used in the manufacture of lithium ion batteries and is actively involved as an extraction solvent in the determination of oil and grease in water by FT-IR.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tetrachloroethylene, Ultrapure | |
°C to 22°C | |
120°C to 121°C | |
C2Cl4 | |
MFCD00000834 | |
1361721 | |
Miscible with alcohol,ether,chloroform,benzene and hexane. Slightly miscible with water. | |
ClC(Cl)=C(Cl)Cl | |
165.82 | |
CHEBI:17300 | |
Spectrophotometric |
127-18-4 | |
1.623 g/mL | |
>99% | |
1.5055 | |
4 x 1 L | |
tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin | |
CYTYCFOTNPOANT-UHFFFAOYSA-N | |
1,1,2,2-tetrachloroethene | |
31373 | |
165.83 | |
Liquid |
Chemical Identifiers
127-18-4 | |
165.82 | |
CYTYCFOTNPOANT-UHFFFAOYSA-N | |
31373 | |
1,1,2,2-tetrachloroethene |
C2Cl4 | |
MFCD00000834 | |
tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin | |
CHEBI:17300 | |
ClC(Cl)=C(Cl)Cl |
Safety and Handling
P201-P202-P260-P264b-P271-P272-P280-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P362-P501c
H315-H317-H319-H336-H350-H373
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 204-825-9
RTECSNumber : KX3850000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only