missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrachloroethylene, Ultrapure, Spectrophotometric Grade, >99%, Alfa Aesar™

Manufacturer:  Alfa Aesar 32437K7

 View more versions of this product

Catalog No. AA32437K7



Specifications

Specifications

Tetrachloroethylene, Ultrapure
4L
Sprectrophotometric
C2Cl4
tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
C(=C(Cl)Cl)(Cl)Cl
165.822
CHEBI:17300
Liquid
1.623 g/mL
127-18-4
>99%
MFCD00000834
CYTYCFOTNPOANT-UHFFFAOYSA-N
1,1,2,2-tetrachloroethene
31373
165.83

Chemical Identifiers

127-18-4
165.822
CYTYCFOTNPOANT-UHFFFAOYSA-N
31373
1,1,2,2-tetrachloroethene
C2Cl4
MFCD00000834
tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
CHEBI:17300
C(=C(Cl)Cl)(Cl)Cl
Safety and Handling

Safety and Handling

EINECSNumber : 204-825-9

SDS Documents