Please login to your online account to display your discounted pricing

Tetrachloroethylene (Technical), Fisher Chemical


Quantity 4L
Packaging Glass Bottle

Manufacturer: fisher scientific company  BPC1824

 View more versions of this product

Catalog No. C182-4

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Formula Weight 165.83
Boiling Point 120°C
Melting Point −22°C
Quantity 4L
DOT Information DOT Class 6.1, : Poison
Physical Form Liquid
Density 1.625g/cm³
Color Colorless
Decomposition Information >150°C
Packaging Glass Bottle
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
Suspect cancer hazard. Irritating to eyes and skin. May cause irritation of respiratory tract. May cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:0
Instability:0
ChemAlert Storage Symbol Blue
MDL Number MFCD00000834
Additional Information Vapor Pressure: 18mbar at 20°C
Synonym Carbon Dichloride
Identification Pass Test
Linear Formula CCl2:CCl2
Molecular Formula C2Cl4
CAS 127-18-4
Chemical Name or Material Tetrachloroethylene

Specifications

Chemical Name or Material Tetrachloroethylene
CAS 127-18-4
Purity Grade Technical Grade
Molecular Formula C2Cl4
Linear Formula CCl2:CCl2
Formula Weight 165.83
MDL Number MFCD00000834
Synonym Carbon Dichloride
Accessories

Accessories for Catalog Number: C1824

Item Mfr. No. Item Description Price Per Unit Quantity  
03-439
Eagle™ Thermoplastics™ Safety Bottle Carriers
fisher scientific
B102
Safety bottle carriers are molded-in handle Each for $217.50
Accessories
 

Emergency Overview
Suspect cancer hazard. Irritating to eyes and skin. May cause irritation of respiratory tract. May cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:0
Instability:0

WARNING!

SDS & Safety Documents