missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrachloroethylene, 99+%, spectrophotometric grade, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 167890025

 View more versions of this product

Catalog No. AC167890025



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tetrachloroethylene
(1cm cell vs water), (1 cm cell vs water) at 0nm, 0.03 max. at 400nm, 0.05 max. at 350nm, 0.1 max. at 305nm, 0.3 max. at 295nm, 1 max. at 290nm
1.6250g/mL
99+%
C2Cl4
MFCD00000834
01, 187
15, 9335
CYTYCFOTNPOANT-UHFFFAOYSA-N
1,1,2,2-tetrachloroethene
31373
165.83
≥99.0% (GC)
127-18-4
>150°C
Authentic
Glass Bottle
Cl2C=CCl2
2.5 L
0.0005% max.
tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
ClC(Cl)=C(Cl)Cl
165.82
CHEBI:17300
18mbar at 20°C
Liquid

Chemical Identifiers

127-18-4
165.82
CYTYCFOTNPOANT-UHFFFAOYSA-N
31373
ClC(Cl)=C(Cl)Cl
C2Cl4
MFCD00000834
tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
CHEBI:17300
Safety and Handling

Safety and Handling

GHS P Statement
Suspected of causing cancer.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause an allergic skin reaction.
May cause drowsiness or dizziness.

GHS P Statement
Use personal protective equipment as required.
Avoid release to the environment.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only