Learn More
Tetrachloroethylene, 99%, extra pure, Thermo Scientific Chemicals
Tetrachloroethylene, 99%, C2Cl4, CAS Number-127-18-4, ankilostin, carbon dichloride, ethene, tetrachloro, ethylene tetrachloride, perc, perchlorethylene, perchloroethylene, tetrachlorethylene, tetrachloroethene, tetrachloroethylene, 2.5L, 0.003% max. (NaOH), 98.5% min. (GC), 01, 187
Supplier: Thermo Scientific Chemicals 138010010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tetrachloroethylene | |
127-18-4 | |
1.6250g/mL | |
Authentic | |
Glass Bottle | |
1.505 | |
MFCD00000834 | |
01, 187 | |
1.625 | |
tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin | |
CYTYCFOTNPOANT-UHFFFAOYSA-N | |
1,1,2,2-tetrachloroethene | |
31373 | |
CHEBI:17300 | |
99% | |
Liquid |
0.003% max. (NaOH) | |
-22.0°C | |
120.0°C to 122.0°C | |
98.5% min. (GC) | |
C2Cl4 | |
Cl2C=CCl2 | |
1 L | |
0.005% max. | |
15, 9335 | |
Solubility in water: 0.15g/L (20°C). Other solubilities: miscible with alcohol, ether, oils, chloroform, and benzene | |
ClC(Cl)=C(Cl)Cl | |
165.82 | |
0.89 mPa.s (20°C) | |
165.83 | |
Extra Pure |
Chemical Identifiers
127-18-4 | |
165.82 | |
CYTYCFOTNPOANT-UHFFFAOYSA-N | |
31373 | |
1,1,2,2-tetrachloroethene |
C2Cl4 | |
MFCD00000834 | |
tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin | |
CHEBI:17300 | |
ClC(Cl)=C(Cl)Cl |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause an allergic skin reaction.
May cause drowsiness or dizziness.
Causes serious eye irritati
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at r
GHS Signal Word: Warning
EINECSNumber : 204-825-9
RUO – Research Use Only