missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrachlorobis(tetrahydrofuran)zirconium(IV), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03955009

 View more versions of this product

Catalog No. AA3955009



Description

Description

It is an important organic intermediate and can be used in agrochemicals, paharmaceuticals and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tetrachlorobis(tetrahydrofuran)zirconium(IV)
21959-01-3
10 g
C8H16Cl4O2Zr
UN2925
Reacts with water.
C1CCOC1.C1CCOC1.Cl[Zr](Cl)(Cl)Cl
377.238
377.25
175°C to 177°C
White
Crystalline
MFCD00145367
tetrachlorobis tetrahydrofuran zirconium, zrcl4 thf 2, oxolane; tetrachlorozirconium, tetrachlorobis tetrahydrofurane zirconium, zirconium tetrachloride tetrahydrofuran complex, zirconium iv chloride tetrahydrofuran complex, bis tetrahydrofuran ; zirconium iv chloride, zirconium iv chloride tetrahydrofuran complex 1:2
VDJJKYYENYIHFF-UHFFFAOYSA-J
oxolane;tetrachlorozirconium
2734040

Chemical Identifiers

21959-01-3
377.238
VDJJKYYENYIHFF-UHFFFAOYSA-J
2734040
C1CCOC1.C1CCOC1.Cl[Zr](Cl)(Cl)Cl
C8H16Cl4O2Zr
MFCD00145367
tetrachlorobis tetrahydrofuran zirconium, zrcl4 thf 2, oxolane; tetrachlorozirconium, tetrachlorobis tetrahydrofurane zirconium, zirconium tetrachloride tetrahydrofuran complex, zirconium iv chloride tetrahydrofuran complex, bis tetrahydrofuran ; zirconium iv chloride, zirconium iv chloride tetrahydrofuran complex 1:2
oxolane;tetrachlorozirconium
Safety and Handling

Safety and Handling

GHS H Statement
H228-H314-H318
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312+H332-H314

DOTInformation : Hazard Class: 8; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only