Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetrachlorobis(tetrahydrofuran)niobium(IV)

$54.85 - $54.85

Chemical Identifiers

Chemical Name or Material Tetrachlorobis(tetrahydrofuran)niobium(IV)
CAS 61247-57-2
Formula Weight 378.94
MDL Number MFCD00013405
UN Number UN2925
Synonym Niobium(IV) chloride - tetrahydrofuran complex
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3946004

  SDS
alfa aesar™
3946004
2g Each for $54.85

AA3946009

  SDS
alfa aesar™
3946009
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 378.94
MDL Number MFCD00013405
Health Hazard 1 Danger
CAS 61247-57-2
DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
Health Hazard 2 GHS H Statement
H228-H314-H318
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flash Point 49° (120°F)
Chemical Name or Material Tetrachlorobis(tetrahydrofuran)niobium(IV)
Synonym Niobium(IV) chloride - tetrahydrofuran complex
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 2g
Sensitivity Moisture Sensitive
Color Yellow-orange to brown
Melting Point 93°C (dec.)
UN Number UN2925
Physical Form Powder
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H228-H314-H318
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.