missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrachloroauric(III) Acid Trihydrate, 99.5%, For Analysis, EMSURE™, MilliporeSigma™

Manufacturer:  MilliporeSigma 1015820005

 View more versions of this product

Catalog No. M1015820005



Description

Description

For analysis
Specifications

Specifications

Tetrachloroauric(III) Acid Trihydrate
99.50%
UN 3260
Gold(III)chloride hydrochloric acid, Gold trichloride hydrochloride
Solid
Fibre case
16961-25-4
AuCl4H·3H2O
Yellow solid
393.83
Odorless
5g

Chemical Identifiers

16961-25-4
AuCl4H·3H2O
Gold(III)chloride hydrochloric acid, Gold trichloride hydrochloride
Safety and Handling

Safety and Handling

H314: Causes severe skin burns and eye damage.
H317: May cause an allergic skin reaction.

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 240-948-4

Documents
Promotions

Promotions